Anti-LYN Mouse Monoclonal Antibody [clone: 338716]

Supplier: R & D Systems
MAB3206
220020-334EA 577.02 USD
220020-334
Anti-LYN Mouse Monoclonal Antibody [clone: 338716]
Antibodies
Type: Primary
Antigen: LYN
Clonality: Monoclonal
Clone: 338716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR