Anti-CCL11 Mouse Monoclonal Antibody [clone: 43915]

Supplier: R & D Systems
MAB3201
220020-332EA 763.57 USD
220020-332
Anti-CCL11 Mouse Monoclonal Antibody [clone: 43915]
Antibodies
Type: Primary
Antigen: CCL11
Clonality: Monoclonal
Clone: 43915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR