Anti-IKBKE Mouse Monoclonal Antibody [clone: 346909]

Supplier: R & D Systems
MAB3199
220020-330EA 594.37 USD
220020-330
Anti-IKBKE Mouse Monoclonal Antibody [clone: 346909]
Antibodies
Type: Primary
Antigen Symbol: IKBKE
Antigen Name: inhibitor of kappaB kinase epsilon
Antigen Synonyms: AW558201, IKK-i, IKKepsilon, Ikki
Clonality: Monoclonal
Clone: 346909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR