Anti-SERPIND1 Mouse Monoclonal Antibody [clone: 373008]

Supplier: R & D Systems
MAB3198
220020-329EA 577.02 USD
220020-329
Anti-SERPIND1 Mouse Monoclonal Antibody [clone: 373008]
Antibodies
Type: Primary
Antigen: SERPIND1
Clonality: Monoclonal
Clone: 373008
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR