Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 362532]

Supplier: R & D Systems
MAB3197
220020-328EA 616.06 USD
220020-328
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 362532]
Antibodies
Simple Western

Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone: 362532
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR