Anti-MAPK6 Mouse Monoclonal Antibody [clone: 335102]

Supplier: R & D Systems
MAB3196
220020-327EA 561.11 USD
220020-327
Anti-MAPK6 Mouse Monoclonal Antibody [clone: 335102]
Antibodies
Type: Primary
Antigen: MAPK6
Clonality: Monoclonal
Clone: 335102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR