Anti-FGF17 Mouse Monoclonal Antibody [clone: 121630]

Supplier: R & D Systems
MAB319
220020-325EA 763.57 USD
220020-325
Anti-FGF17 Mouse Monoclonal Antibody [clone: 121630]
Antibodies
Type: Primary
Antigen Symbol: FGF17
Antigen Name: fibroblast growth factor 17
Antigen Synonyms: FGF-13, FGF-17, HH20
Clonality: Monoclonal
Clone: 121630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR