Anti-F3 Rat Monoclonal Antibody [clone: 355220]

Supplier: R & D Systems
MAB3178
220020-324EA 577.02 USD
220020-324
Anti-F3 Rat Monoclonal Antibody [clone: 355220]
Antibodies
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: 355220
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2C
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR