Anti-BDNF Mouse Monoclonal Antibody [clone: 584412]

Supplier: R & D Systems
MAB31751
102885-550EA 594.37 USD
102885-550
Anti-BDNF Mouse Monoclonal Antibody [clone: 584412]
Antibodies
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone: 584412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR