Anti-BDNF Mouse Monoclonal Antibody [clone: 584421]

Supplier: R & D Systems
MAB3175
102885-548EA 577.02 USD
102885-548
Anti-BDNF Mouse Monoclonal Antibody [clone: 584421]
Antibodies
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone: 584421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR