Anti-MPO Mouse Monoclonal Antibody [clone: 392105]

Supplier: R & D Systems
MAB3174
220020-322EA 655.11 USD
220020-322
Anti-MPO Mouse Monoclonal Antibody [clone: 392105]
Antibodies
Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 392105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR