Anti-HMOX2 Mouse Monoclonal Antibody [clone: 322913]

Supplier: R & D Systems
MAB3170
220020-320EA 577.02 USD
220020-320
Anti-HMOX2 Mouse Monoclonal Antibody [clone: 322913]
Antibodies
Type: Primary
Antigen: HMOX2
Clonality: Monoclonal
Clone: 322913
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR