Anti-C1QTNF5 Mouse Monoclonal Antibody [clone: 332923]

Supplier: R & D Systems
MAB3167
220020-318EA 633.42 USD
220020-318
Anti-C1QTNF5 Mouse Monoclonal Antibody [clone: 332923]
Antibodies
Type: Primary
Antigen: C1QTNF5
Clonality: Monoclonal
Clone: 332923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR