Anti-CK8 Mouse Monoclonal Antibody [clone: LP3K]

Supplier: R & D Systems
MAB3165
220020-317EA 594.37 USD
220020-317
Anti-CK8 Mouse Monoclonal Antibody [clone: LP3K]
Antibodies
Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: LP3K
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR