Anti-CK14 Mouse Monoclonal Antibody [clone: LL001]

Supplier: R & D Systems
MAB3164
220020-316EA 594.37 USD
220020-316
Anti-CK14 Mouse Monoclonal Antibody [clone: LL001]
Antibodies
Type: Primary
Antigen: CK14
Clonality: Monoclonal
Clone: LL001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR