Anti-IL16 Mouse Monoclonal Antibody [clone: 70720]

Supplier: R & D Systems
MAB3161
220020-315EA 594.37 USD
220020-315
Anti-IL16 Mouse Monoclonal Antibody [clone: 70720]
Antibodies
Type: Primary
Antigen: IL16
Clonality: Monoclonal
Clone: 70720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR