Anti-WNT9A Mouse Monoclonal Antibody [clone: 326707]

Supplier: R & D Systems
MAB31571
220020-313EA 594.37 USD
220020-313
Anti-WNT9A Mouse Monoclonal Antibody [clone: 326707]
Antibodies
Type: Primary
Antigen: WNT9A
Clonality: Monoclonal
Clone: 326707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR