Anti-SORT1 Mouse Monoclonal Antibody [clone: 334708]

Supplier: R & D Systems
MAB3154
220020-310EA 594.37 USD
220020-310
Anti-SORT1 Mouse Monoclonal Antibody [clone: 334708]
Antibodies
Simple Western

Type: Primary
Antigen: SORT1
Clonality: Monoclonal
Clone: 334708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR