Anti-FABP4 Rat Monoclonal Antibody [clone: 804914]

Supplier: R & D Systems
MAB3150
102885-546EA 594.37 USD
102885-546
Anti-FABP4 Rat Monoclonal Antibody [clone: 804914]
Antibodies
Type: Primary
Antigen Symbol: FABP4
Antigen Name: fatty acid binding protein 4, adipocyte
Antigen Synonyms: A-FABP, AFABP, ALBP, HEL-S-104, aP2
Clonality: Monoclonal
Clone: 804914
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR