Anti-IL1A Mouse Monoclonal Antibody [clone: 85733]

Supplier: R & D Systems
MAB315
220020-309EA 772.25 USD
220020-309
Anti-IL1A Mouse Monoclonal Antibody [clone: 85733]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 85733
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR