Anti-OPTC Rat Monoclonal Antibody [clone: 361328]

Supplier: R & D Systems
MAB3148
220020-308EA 561.11 USD
220020-308
Anti-OPTC Rat Monoclonal Antibody [clone: 361328]
Antibodies
Type: Primary
Antigen: OPTC
Clonality: Monoclonal
Clone: 361328
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR