Anti-C1QTNF1 Mouse Monoclonal Antibody [clone: 395526]

Supplier: R & D Systems
MAB3145
220020-306EA 577.02 USD
220020-306
Anti-C1QTNF1 Mouse Monoclonal Antibody [clone: 395526]
Antibodies
Type: Primary
Antigen: C1QTNF1
Clonality: Monoclonal
Clone: 395526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR