Anti-MEPE Mouse Monoclonal Antibody [clone: 341611]

Supplier: R & D Systems
MAB3140
220020-305EA 566.89 USD
220020-305
Anti-MEPE Mouse Monoclonal Antibody [clone: 341611]
Antibodies
Type: Primary
Antigen: MEPE
Clonality: Monoclonal
Clone: 341611
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR