Anti-COMP Mouse Monoclonal Antibody [clone: 2127-F5]

Supplier: R & D Systems
MAB31341
220020-304EA 577.02 USD
220020-304
Anti-COMP Mouse Monoclonal Antibody [clone: 2127-F5]
Antibodies
Type: Primary
Antigen: COMP
Clonality: Monoclonal
Clone: 2127-F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR