Anti-COMP Mouse Monoclonal Antibody [clone: 360215]

Supplier: R & D Systems
MAB3134
220020-303EA 577.02 USD
220020-303
Anti-COMP Mouse Monoclonal Antibody [clone: 360215]
Antibodies
Type: Primary
Antigen: COMP
Clonality: Monoclonal
Clone: 360215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR