Anti-COLEC12 Rat Monoclonal Antibody [clone: 350611]

Supplier: R & D Systems
MAB3130
220020-299EA 561.11 USD
220020-299
Anti-COLEC12 Rat Monoclonal Antibody [clone: 350611]
Antibodies
Type: Primary
Antigen: COLEC12
Clonality: Monoclonal
Clone: 350611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR