Anti-TEK Mouse Monoclonal Antibody [clone: 83711]

Supplier: R & D Systems
MAB313
220020-298EA 772.25 USD
220020-298
Anti-TEK Mouse Monoclonal Antibody [clone: 83711]
Antibodies
Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 83711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR