Anti-APLP1 Mouse Monoclonal Antibody [clone: 3548]

Supplier: R & D Systems
MAB3129
220020-297EA 594.37 USD
220020-297
Anti-APLP1 Mouse Monoclonal Antibody [clone: 3548]
Antibodies
Type: Primary
Antigen: APLP1
Clonality: Monoclonal
Clone: 3548
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR