Anti-NPPC Rat Monoclonal Antibody [clone: 462608]

Supplier: R & D Systems
MAB31271
220020-296EA 594.37 USD
220020-296
Anti-NPPC Rat Monoclonal Antibody [clone: 462608]
Antibodies
Type: Primary
Antigen: NPPC
Clonality: Monoclonal
Clone: 462608
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR