Anti-DPPA5 Mouse Monoclonal Antibody [clone: 319606]

Supplier: R & D Systems
MAB3125
220020-294EA 561.11 USD
220020-294
Anti-DPPA5 Mouse Monoclonal Antibody [clone: 319606]
Antibodies
Type: Primary
Antigen: DPPA5
Clonality: Monoclonal
Clone: 319606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR