Anti-SERPINI1 Rat Monoclonal Antibody [clone: 437016]

Supplier: R & D Systems
MAB3124
220020-293EA 561.11 USD
220020-293
Anti-SERPINI1 Rat Monoclonal Antibody [clone: 437016]
Antibodies
Type: Primary
Antigen: SERPINI1
Clonality: Monoclonal
Clone: 437016
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR