Anti-TFR2 Mouse Monoclonal Antibody [clone: 353816]

Supplier: R & D Systems
MAB3120
220020-292EA 623.29 USD
220020-292
Anti-TFR2 Mouse Monoclonal Antibody [clone: 353816]
Antibodies
Simple Western

Type: Primary
Antigen: TFR2
Clonality: Monoclonal
Clone: 353816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR