Anti-PVRL4 Rat Monoclonal Antibody [clone: 356704]

Supplier: R & D Systems
MAB3116
220020-291EA 600.15 USD
220020-291
Anti-PVRL4 Rat Monoclonal Antibody [clone: 356704]
Antibodies
Type: Primary
Antigen: PVRL4
Clonality: Monoclonal
Clone: 356704
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR