Anti-EPYC Mouse Monoclonal Antibody [clone: 329707]

Supplier: R & D Systems
MAB3110
220020-289EA 561.11 USD
220020-289
Anti-EPYC Mouse Monoclonal Antibody [clone: 329707]
Antibodies
Type: Primary
Antigen: EPYC
Clonality: Monoclonal
Clone: 329707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR