Anti-MYD88 Rat Monoclonal Antibody [clone: 316902]

Supplier: R & D Systems
MAB3109
220020-288EA 594.37 USD
220020-288
Anti-MYD88 Rat Monoclonal Antibody [clone: 316902]
Antibodies
Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 316902
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR