Anti-NEFH Mouse Monoclonal Antibody [clone: 321721]

Supplier: R & D Systems
MAB3108
220020-287EA 577.02 USD
220020-287
Anti-NEFH Mouse Monoclonal Antibody [clone: 321721]
Antibodies
Type: Primary
Antigen Symbol: NEFH
Antigen Name: neurofilament, heavy polypeptide
Antigen Synonyms: NFH
Clonality: Monoclonal
Clone: 321721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR