Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 861812]

Supplier: R & D Systems
MAB3100
102885-540EA 577.02 USD
102885-540
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 861812]
Antibodies
Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 861812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR