Anti-RTN4 Mouse Monoclonal Antibody [clone: 391401]

Supplier: R & D Systems
MAB30981
220020-283EA 577.02 USD
220020-283
Anti-RTN4 Mouse Monoclonal Antibody [clone: 391401]
Antibodies
Type: Primary
Antigen: RTN4
Clonality: Monoclonal
Clone: 391401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR