Anti-RTN4 Mouse Monoclonal Antibody [clone: 387521]

Supplier: R & D Systems
MAB3098
102885-538EA 594.37 USD
102885-538
Anti-RTN4 Mouse Monoclonal Antibody [clone: 387521]
Antibodies
Type: Primary
Antigen: RTN4
Clonality: Monoclonal
Clone: 387521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR