Anti-FBLN5 Mouse Monoclonal Antibody [clone: 293904]

Supplier: R & D Systems
MAB3095
220020-282EA 594.37 USD
220020-282
Anti-FBLN5 Mouse Monoclonal Antibody [clone: 293904]
Antibodies
Type: Primary
Antigen Symbol: FBLN5
Antigen Name: fibulin 5
Antigen Synonyms: ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, UP50
Clonality: Monoclonal
Clone: 293904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR