Anti-EGFL7 Rat Monoclonal Antibody [clone: 362907]

Supplier: R & D Systems
MAB3089
220020-281EA 577.02 USD
220020-281
Anti-EGFL7 Rat Monoclonal Antibody [clone: 362907]
Antibodies
Type: Primary
Antigen: EGFL7
Clonality: Monoclonal
Clone: 362907
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR