Anti-MEA-1 Rat Monoclonal Antibody [clone: 364605]

Supplier: R & D Systems
MAB3088
220020-280EA 561.11 USD
220020-280
Anti-MEA-1 Rat Monoclonal Antibody [clone: 364605]
Antibodies
Type: Primary
Antigen Symbol: MEA-1
Antigen Name:
Antigen Synonyms: HYS
Clonality: Monoclonal
Clone: 364605
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR