Anti-CRHBP Rat Monoclonal Antibody [clone: 356803]

Supplier: R & D Systems
MAB3087
220020-279EA 561.11 USD
220020-279
Anti-CRHBP Rat Monoclonal Antibody [clone: 356803]
Antibodies
Type: Primary
Antigen: CRHBP
Clonality: Monoclonal
Clone: 356803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR