Anti-LINGO1 Mouse Monoclonal Antibody [clone: 332237]

Supplier: R & D Systems
MAB30861
220020-278EA 594.37 USD
220020-278
Anti-LINGO1 Mouse Monoclonal Antibody [clone: 332237]
Antibodies
Type: Primary
Antigen: LINGO1
Clonality: Monoclonal
Clone: 332237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR