Anti-LILRA1 Mouse Monoclonal Antibody [clone: 586326]

Supplier: R & D Systems
MAB30851
220020-276EA 655.11 USD
220020-276
Anti-LILRA1 Mouse Monoclonal Antibody [clone: 586326]
Antibodies
Type: Primary
Antigen: LILRA1
Clonality: Monoclonal
Clone: 586326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR