Anti-LILRA1 Mouse Monoclonal Antibody [clone: 356316]

Supplier: R & D Systems
MAB3085
220020-275EA 642.09 USD
220020-275
Anti-LILRA1 Mouse Monoclonal Antibody [clone: 356316]
Antibodies
Type: Primary
Antigen: LILRA1
Clonality: Monoclonal
Clone: 356316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR