Anti-FAM3C Mouse Monoclonal Antibody [clone: 438618]

Supplier: R & D Systems
MAB3084
220020-274EA 561.11 USD
220020-274
Anti-FAM3C Mouse Monoclonal Antibody [clone: 438618]
Antibodies
Type: Primary
Antigen Symbol: FAM3C
Antigen Name: family with sequence similarity 3, member C
Antigen Synonyms: GS3786, ILEI
Clonality: Monoclonal
Clone: 438618
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR