Anti-GPA33 Rat Monoclonal Antibody [clone: 402104]

Supplier: R & D Systems
MAB3080
220020-273EA 600.15 USD
220020-273
Anti-GPA33 Rat Monoclonal Antibody [clone: 402104]
Antibodies
Type: Primary
Antigen Symbol: GPA33
Antigen Name: glycoprotein A33 (transmembrane)
Antigen Synonyms: A33
Clonality: Monoclonal
Clone: 402104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR