Anti-FABP5 Rat Monoclonal Antibody [clone: 311215]

Supplier: R & D Systems
MAB3077
220020-270EA 623.29 USD
220020-270
Anti-FABP5 Rat Monoclonal Antibody [clone: 311215]
Antibodies
Type: Primary
Antigen Symbol: FABP5
Antigen Name: fatty acid binding protein 5 (psoriasis-associated)
Antigen Synonyms: E-FABP, EFABP, KFABP, PA-FABP, PAFABP
Clonality: Monoclonal
Clone: 311215
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR