Anti-EPOR Mouse Monoclonal Antibody [clone: 690748]

Supplier: R & D Systems
MAB3072
102885-536EA 594.37 USD
102885-536
Anti-EPOR Mouse Monoclonal Antibody [clone: 690748]
Antibodies
Type: Primary
Antigen: EPOR
Clonality: Monoclonal
Clone: 690748
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR